Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exo5
Entrez Gene ID
313563
Gene Name
exonuclease 5
Gene Aliases
Dem1, RGD1309802
Chromosome Location
Chr.5: 139849930 - 139853247 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107973.1 NP_001101443.1 1 - 2 517 63

View Full Product Details