Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map7d1
Entrez Gene ID
681287
Gene Name
MAP7 domain containing 1
Gene Aliases
Mtap7d1
Chromosome Location
Chr.5: 144250416 - 144274981 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109438.1 NP_001102908.1 16 - 17 2505 139
XM_006238901.2 XP_006238963.1 15 - 16 2581 139
XM_006238902.2 XP_006238964.1 15 - 16 2572 139
XM_006238903.2 XP_006238965.1 14 - 15 2470 139
XM_006238904.2 XP_006238966.1 14 - 15 2461 139
XM_006238906.2 XP_006238968.1 15 - 16 2365 139
GenBank mRNA FQ216284.1 - 6 - 7 650 139

View Full Product Details