Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf19b
Entrez Gene ID
313806
Gene Name
ring finger protein 19B
Gene Aliases
Ibrdc3
Chromosome Location
Chr.5: 147246032 - 147270892 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108003.1 NP_001101473.1 4 - 5 623 117
XM_008764147.1 XP_008762369.1 6 - 7 1190 117

View Full Product Details