Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tardbp
Entrez Gene ID
298648
Gene Name
TAR DNA binding protein
Gene Aliases
-
Chromosome Location
Chr.5: 165432089 - 165442112 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011979.2 NP_001011979.1 2 - 3 335 103
XM_006239382.2 XP_006239444.1 2 - 3 388 103
XM_006239384.2 XP_006239446.1 2 - 3 365 103
XM_006239385.2 XP_006239447.1 2 - 3 366 103
XM_006239386.2 XP_006239448.1 2 - 3 370 103
XM_008764234.1 XP_008762456.1 2 - 3 367 103
XM_008764235.1 XP_008762457.1 2 - 3 365 103
XM_008764236.1 XP_008762458.1 2 - 3 365 103
GenBank mRNA AI502405.1 - 2 - 3 346 103
BC083752.1 - 2 - 3 324 103

View Full Product Details