The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011979.2 | NP_001011979.1 | 4 - 5 | 644 | 78 |
XM_006239382.2 | XP_006239444.1 | 4 - 5 | 697 | 78 | |
XM_006239384.2 | XP_006239446.1 | 4 - 5 | 674 | 78 | |
XM_006239385.2 | XP_006239447.1 | 4 - 5 | 675 | 78 | |
XM_006239386.2 | XP_006239448.1 | 4 - 5 | 679 | 78 | |
XM_008764234.1 | XP_008762456.1 | 4 - 5 | 676 | 78 | |
XM_008764235.1 | XP_008762457.1 | 4 - 5 | 674 | 78 | |
XM_008764236.1 | XP_008762458.1 | 4 - 5 | 674 | 78 | |
GenBank mRNA | BC083752.1 | - | 4 - 5 | 633 | 78 |