Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd5
Entrez Gene ID
301039
Gene Name
sushi domain containing 5
Gene Aliases
RGD1561132
Chromosome Location
Chr.8: 122331488 - 122370672 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_236674.7 XP_236674.5 4 - 5 687 87

View Full Product Details