Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnpc3
Entrez Gene ID
691538
Gene Name
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 216481457 - 216510054 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100810.1 NP_001094280.1 2 - 3 291 122
XM_006233211.2 XP_006233273.1 2 - 3 669 122
XM_008761424.1 XP_008759646.1 2 - 3 669 122
XM_008761425.1 XP_008759647.1 2 - 3 670 122
GenBank mRNA BC166571.1 - 2 - 3 291 122

View Full Product Details