Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddit4l
Entrez Gene ID
100363484
Gene Name
DNA-damage-inducible transcript 4-like
Gene Aliases
-
Chromosome Location
Chr.2: 242882229 - 242885088 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729121.4 XP_002729167.1 1 - 2 232 68

View Full Product Details