The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126120.1 | NP_001119592.1 | 8 - 9 | 1117 | 97 |
GenBank mRNA | BC083724.1 | - | 8 - 9 | 1117 | 97 |
FQ213560.1 | - | 8 - 9 | 1163 | 97 |