Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpf38b
Entrez Gene ID
499691
Gene Name
pre-mRNA processing factor 38B
Gene Aliases
-
Chromosome Location
Chr.2: 211706245 - 211715271 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024305.2 NP_001019476.1 1 - 2 560 70
GenBank mRNA BC079264.1 - 1 - 2 509 70
FQ210223.1 - 1 - 2 540 70

View Full Product Details