Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prpf38b
Entrez Gene ID
499691
Gene Name
pre-mRNA processing factor 38B
Gene Aliases
-
Chromosome Location
Chr.2: 211706245 - 211715271 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024305.2 NP_001019476.1 2 - 3 630 137
GenBank mRNA BC079264.1 - 2 - 3 579 137
FQ210223.1 - 2 - 3 610 137

View Full Product Details