The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012183.2 | NP_001012183.1 | 8 - 9 | 900 | 61 |
XM_006233520.2 | XP_006233582.1 | 9 - 10 | 1136 | 61 | |
GenBank mRNA | BC078927.1 | - | 8 - 9 | 902 | 61 |