Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dapp1
Entrez Gene ID
362046
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides
Gene Aliases
-
Chromosome Location
Chr.2: 243175346 - 243224883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108568.1 NP_001102038.1 6 - 7 710 79

View Full Product Details