The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199494.1 | NP_955788.1 | 2 - 3 | 251 | 86 |
XM_006233519.2 | XP_006233581.1 | 2 - 3 | 277 | 86 | |
GenBank mRNA | AJ276066.1 | - | 2 - 3 | 286 | 86 |
BC088089.1 | - | 2 - 3 | 253 | 86 | |
BC168244.1 | - | 2 - 3 | 234 | 86 |