Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc7
Entrez Gene ID
310787
Gene Name
fibronectin type III domain containing 7
Gene Aliases
RGD1309189
Chromosome Location
Chr.2: 211655482 - 211690799 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107717.1 NP_001101187.1 2 - 3 383 118
XM_006233159.2 XP_006233221.1 3 - 4 391 118
XM_006233160.2 XP_006233222.1 3 - 4 384 118

View Full Product Details