Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gstm6
Entrez Gene ID
499688
Gene Name
glutathione S-transferase, mu 6
Gene Aliases
RGD1563391
Chromosome Location
Chr.2: 210733594 - 210738368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109192.1 NP_001102662.1 1 - 2 146 101
GenBank mRNA BC158681.1 - 1 - 2 156 101

View Full Product Details