Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfi
Entrez Gene ID
79126
Gene Name
complement factor I
Gene Aliases
FI
Chromosome Location
Chr.2: 235264149 - 235305779 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024157.1 NP_077071.1 12 - 13 1489 78
XM_008761526.1 XP_008759748.1 13 - 14 1827 78
GenBank mRNA BC089798.1 - 12 - 13 1494 78
Y18965.1 - 12 - 13 1489 78

View Full Product Details