Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac3
Entrez Gene ID
29758
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3
Gene Aliases
Siat7c
Chromosome Location
Chr.2: 259379653 - 259918813 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019123.1 NP_061996.1 1 - 2 246 83
GenBank mRNA BC072501.1 - 1 - 2 83 83
L29554.1 - 1 - 2 246 83

View Full Product Details