Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Palmd
Entrez Gene ID
310811
Gene Name
palmdelphin
Gene Aliases
-
Chromosome Location
Chr.2: 220481292 - 220536004 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025688.1 NP_001020859.1 4 - 5 1871 85
GenBank mRNA BC098755.1 - 4 - 5 1871 85

View Full Product Details