Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syde2
Entrez Gene ID
308021
Gene Name
synapse defective 1, Rho GTPase, homolog 2 (C. elegans)
Gene Aliases
RGD1564206, Syde2-ps1
Chromosome Location
Chr.2: 251861902 - 251895524 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749414.3 XP_003749462.1 1 - 2 1881 80

View Full Product Details