The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003749414.3 | XP_003749462.1 | 5 - 6 | 3955 | 67 |
GenBank mRNA | FQ218152.1 | - | 3 - 4 | 460 | 67 |