The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004090.2 | NP_001004090.2 | 2 - 3 | 140 | 59 |
GenBank mRNA | AF540877.1 | - | 2 - 3 | 166 | 59 |
BC100135.1 | - | 2 - 3 | 140 | 59 |