Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
65026
Gene Name
RWD domain containing 3
Gene Aliases
Rsume, X2cr1
Chromosome Location
Chr.2: 224710351 - 224723129 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128152.1 NP_001121624.1 2 - 3 591 123

View Full Product Details