Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ostc
Entrez Gene ID
362040
Gene Name
oligosaccharyltransferase complex subunit (non-catalytic)
Gene Aliases
Dc2, RGD1560708
Chromosome Location
Chr.2: 235816328 - 235831651 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108566.1 NP_001102036.1 2 - 3 277 74
GenBank mRNA BC159425.1 - 2 - 3 263 74
FQ219812.1 - 2 - 3 283 74
FQ219920.1 - 2 - 3 291 74
FQ220255.1 - 2 - 3 259 74
FQ230162.1 - 2 - 3 259 74
FQ234814.1 - 2 - 3 239 74

View Full Product Details