Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rabggtb
Entrez Gene ID
25533
Gene Name
Rab geranylgeranyltransferase, beta subunit
Gene Aliases
-
Chromosome Location
Chr.2: 260110309 - 260115574 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138708.2 NP_619715.1 3 - 4 423 99
GenBank mRNA BC060536.1 - - 447 99
BC078683.1 - 3 - 4 412 99
FQ214773.1 - - 427 99
FQ216744.1 - - 427 99
FQ224712.1 - - 426 99
FQ229109.1 - - 427 99
L10416.1 - 3 - 4 423 99
S62097.1 - 3 - 4 423 99

View Full Product Details