Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tacr3
Entrez Gene ID
24808
Gene Name
tachykinin receptor 3
Gene Aliases
Nkr, Nmkr, Tac3r
Chromosome Location
Chr.2: 240021152 - 240118971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017053.1 NP_058749.1 4 - 5 1272 64
GenBank mRNA J05189.1 - 4 - 5 1272 64

View Full Product Details