The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106203.2 | NP_001099673.1 | 9 - 10 | 1383 | 72 |
GenBank mRNA | BC161957.1 | - | 9 - 10 | 1383 | 72 |