Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mro
Entrez Gene ID
361348
Gene Name
maestro
Gene Aliases
RGD1559803
Chromosome Location
Chr.18: 69834235 - 69850164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254938.2 XP_006255000.1 3 - 4 506 117

View Full Product Details