Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lipg
Entrez Gene ID
291437
Gene Name
lipase, endothelial
Gene Aliases
lipase
Chromosome Location
Chr.18: 70903528 - 70924434 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012741.1 NP_001012759.1 1 - 2 93 71
GenBank mRNA AY916123.1 - 1 - 2 93 71

View Full Product Details