The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012741.1 | NP_001012759.1 | 8 - 9 | 1383 | 163 |
GenBank mRNA | AY916123.1 | - | 8 - 9 | 1383 | 163 |