Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cybb
Entrez Gene ID
66021
Gene Name
cytochrome b-245, beta polypeptide
Gene Aliases
Gp91-phox, Nox2
Chromosome Location
Chr.X: 14578330 - 14610049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023965.1 NP_076455.1 10 - 11 1319 87
GenBank mRNA AF298656.3 - 10 - 11 1319 87
AJ295950.1 - 10 - 11 1319 87
FQ217794.1 - - 1388 87
FQ231233.1 - - 1371 87

View Full Product Details