Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qki
Entrez Gene ID
100910212
Gene Name
QKI, KH domain containing, RNA binding
Gene Aliases
-
Chromosome Location
Chr.1: 50983133 - 51061103 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758780.1 XP_008757002.1 - 1306 92

View Full Product Details