Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnah5
Entrez Gene ID
294854
Gene Name
dynein, axonemal, heavy chain 5
Gene Aliases
DLP5, Dnahc5, Dnahc8
Chromosome Location
Chr.2: 80947730 - 81143997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760824.1 XP_008759046.1 - 1731 64

View Full Product Details