Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cabs1
Entrez Gene ID
64029
Gene Name
calcium-binding protein, spermatid-specific 1
Gene Aliases
Clph, Rsd6
Chromosome Location
Chr.14: 21746294 - 21748356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022263.2 NP_071599.2 1 - 2 1318 148
GenBank mRNA AF271155.1 - 1 - 2 1014 148
BC079062.1 - 1 - 2 1318 148

View Full Product Details