Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csn2
Entrez Gene ID
29173
Gene Name
casein beta
Gene Aliases
Csnb
Chromosome Location
Chr.14: 21972274 - 21979506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AA964718.1 - 1 - 2 193

View Full Product Details