The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044274.1 | NP_001037739.1 | 10 - 11 | 859 | 152 |
GenBank mRNA | DQ198380.1 | - | 10 - 11 | 859 | 152 |