Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epb41l2
Entrez Gene ID
309557
Gene Name
erythrocyte membrane protein band 4.1-like 2
Gene Aliases
Epb4.1l2, RGD1563977
Chromosome Location
Chr.1: 20881820 - 21067713 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006227709.1 XP_006227771.1 7 - 8 1328 99
XM_008758646.1 XP_008756868.1 7 - 8 1510 99
XM_008758647.1 XP_008756869.1 7 - 8 1626 99
XM_008758648.1 XP_008756870.1 7 - 8 1231 99
XM_008758649.1 XP_008756871.1 8 - 9 1466 99
XM_008758650.1 XP_008756872.1 7 - 8 4313 99
GenBank mRNA FQ212032.1 - 8 - 9 1424 99

View Full Product Details