Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adamts15
Entrez Gene ID
300474
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif, 15
Gene Aliases
-
Chromosome Location
Chr.8: 31977001 - 32000378 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106810.1 NP_001100280.1 6 - 7 2223 70

View Full Product Details