Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpy19l2
Entrez Gene ID
300461
Gene Name
dpy-19-like 2 (C. elegans)
Gene Aliases
Dyp19l2, RGD1564311
Chromosome Location
Chr.8: 25711076 - 25829595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242722.2 XP_006242784.1 2 - 3 562 94
XM_235972.8 XP_235972.6 2 - 3 562 94

View Full Product Details