Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hyls1
Entrez Gene ID
680262
Gene Name
hydrolethalus syndrome 1
Gene Aliases
-
Chromosome Location
Chr.8: 36763470 - 36772671 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729925.3 XP_002729971.1 1 - 2 679 103

View Full Product Details