Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Opcml
Entrez Gene ID
116597
Gene Name
opioid binding protein/cell adhesion molecule-like
Gene Aliases
-
Chromosome Location
Chr.8: 29453178 - 29967300 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053848.1 NP_446300.1 6 - 7 1409 64
XM_008766041.1 XP_008764263.1 4 - 5 567 64
GenBank mRNA M88709.1 - - 1559 64
M88710.1 - 6 - 7 1240 64
M88711.1 - 6 - 7 1409 64

View Full Product Details