Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Herpud2
Entrez Gene ID
300463
Gene Name
HERPUD family member 2
Gene Aliases
RGD1307343
Chromosome Location
Chr.8: 26266571 - 26311528 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024988.1 NP_001020159.1 1 - 2 487 118
XM_006242723.2 XP_006242785.1 1 - 2 665 118
GenBank mRNA BC081861.1 - 1 - 2 487 118

View Full Product Details