Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thyn1
Entrez Gene ID
300470
Gene Name
thymocyte nuclear protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 28045093 - 28053974 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007661.2 NP_001007662.1 5 - 6 534 98
GenBank mRNA BC064031.1 - 5 - 6 538 98
FQ213324.1 - 4 - 5 683 98

View Full Product Details