Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shroom4
Entrez Gene ID
317391
Gene Name
shroom family member 4
Gene Aliases
RGD1563434
Chromosome Location
Chr.X: 16719818 - 16929829 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191730.1 NP_001178659.1 5 - 6 2947 80
XM_006256763.2 XP_006256825.1 4 - 5 3024 80

View Full Product Details