Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plcl2
Entrez Gene ID
301173
Gene Name
phospholipase C-like 2
Gene Aliases
-
Chromosome Location
Chr.9: 1782536 - 1984654 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106880.1 NP_001100350.1 6 - 7 3172 89

View Full Product Details