Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb1
Entrez Gene ID
316164
Gene Name
SATB homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 2181132 - 2277949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012129.2 NP_001012129.1 9 - 10 1838 134
XM_006244219.2 XP_006244281.1 9 - 10 1953 134
XM_006244220.2 XP_006244282.1 9 - 10 3867 134
XM_006244221.2 XP_006244283.1 9 - 10 1835 134
XM_008766760.1 XP_008764982.1 8 - 9 2498 134
XM_008766761.1 XP_008764983.1 9 - 10 2467 134
XM_008766762.1 XP_008764984.1 10 - 11 1872 134
XM_008766763.1 XP_008764985.1 10 - 11 4965 134
XM_008766764.1 XP_008764986.1 8 - 9 2498 134
GenBank mRNA BC085814.1 - 9 - 10 1839 134

View Full Product Details