Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad1
Entrez Gene ID
294800
Gene Name
RAD1 checkpoint DNA exonuclease
Gene Aliases
-
Chromosome Location
Chr.2: 60461587 - 60469646 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106419.1 NP_001099889.1 5 - 6 979 126
XM_006232058.2 XP_006232120.1 5 - 6 897 126
XM_006232059.2 XP_006232121.1 5 - 6 861 126

View Full Product Details