Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly6g5c
Entrez Gene ID
294245
Gene Name
lymphocyte antigen 6 complex, locus G5C
Gene Aliases
re8
Chromosome Location
Chr.20: 5100415 - 5104494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198739.3 NP_942034.3 1 - 2 122 104
GenBank mRNA AJ515240.1 - 1 - 2 104

View Full Product Details