Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsph3
Entrez Gene ID
361476
Gene Name
radial spoke 3 homolog (Chlamydomonas)
Gene Aliases
Rshl2
Chromosome Location
Chr.1: 47412166 - 47478925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748694.3 XP_003748742.1 5 - 6 2718 59
XM_006227909.2 XP_006227971.1 5 - 6 2718 59
XM_006227910.2 XP_006227972.1 5 - 6 2718 59
XM_006227911.2 XP_006227973.1 5 - 6 2718 59
XM_008758741.1 XP_008756963.1 5 - 6 2718 59
XM_008758742.1 XP_008756964.1 5 - 6 2718 59
GenBank mRNA BC169129.1 - 3 - 4 468 59

View Full Product Details