Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdss1
Entrez Gene ID
100364990
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
-
Chromosome Location
Chr.17: 90033432 - 90072509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751734.3 XP_003751782.2 4 - 5 1569 65
XM_008771923.1 XP_008770145.1 4 - 5 1609 65
GenBank mRNA BC158695.1 - 4 - 5 449 65
FQ220359.1 - 1 - 2 39 65

View Full Product Details